Learn More
BPS Laminar flow cabinet, BPS eco air H, 6ft horizontal
Brand:  BPS O00002600000
Product Code. 15454832
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Air H horizontal laminar flow cabinets offering product protection. Epoxy powder coated exterior with option for AISI 304L stainless steel exterior (request special model). Stainless steel work surface or non-reflective melaminic panel available as option, safety glass side windows with holes for installation of service fixtures/taps, frontal closure shutter on demand, membrane keyboard front panel with touch sensitive buttons. DEHS 100% port, one electrical socket for all cabinet sizes included as standard.
  • External dimensions: W1860 x H1195 x D925mm
  • H14 HEPA filters
  • 304 stainless steel work surface
  • Safety glass side windows
  • Microprocessor controls
  • Night set-back option for energy saving
  • One electrical socket included as standard