missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lamin B2 Recombinant Rabbit Monoclonal Antibody (23GB2605), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553499
This item is not returnable.
View return policy
Description
Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but dominant genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
Specifications
| Lamin B2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LMNB2 | |
| EPM9; fc15d06; im:7142331; LAMB2; lamin; lamin A/C; lamin B2; lamin B2 S homeolog; lamin B3; lamin-B2; Lamin-L(II); LMN2; LMNA; lmnb2; lmnb2.S; lmnb2-a; lmnb2-b; MGC2721; RGD1563803; unnamed protein product; wu:fb94e05; wu:fb95e12; wu:fc15d06; wu:fc49h03; XELAEV_18009631mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84823 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2605 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q03252 | |
| LMNB2 | |
| A synthesized peptide derived from human Lamin B2 (570-620AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction