missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B1 Mouse anti-Human, Mouse, Rat, Clone: 3C10G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66095-1-IG-150UL
This item is not returnable.
View return policy
Description
Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2.Specifications
| Lamin B1 | |
| Monoclonal | |
| 1.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14733, P20700, P70615 | |
| LMNB1 | |
| Lamin B1 Fusion Protein Ag20522 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 3C10G12 | |
| Unconjugated | |
| LMNB1 | |
| ADLD, lamin B1, LMN, LMN2, LMNB, LMNB1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 116685, 16906, 4001 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction