missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lamin A/C Recombinant Rabbit Monoclonal Antibody (4L8Q0)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535284
This item is not returnable.
View return policy
Description
Immunogen sequence: SSHSSQTQGG GSVTKKRKLE STESRSSFSQ HARTSGRVAV EEVDEEGKFV RLRNKSNEDQ SMGNWQIKRQ NGDDPLLTYR FPPKFTLKAG QVVTIWAAGA GATHSPPTDL VWKAQNTWGC GNSLRTALIN STGEEVAMRK LVRSVTVVED DEDEDGDDLL HHHHGSHCSS.
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).
Specifications
| Lamin A/C | |
| Recombinant Monoclonal | |
| 0.24 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P02545, P48678, P48679 | |
| LMNA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 403-572 of human Lamin A/C (NP_7338211). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 4L8Q0 | |
| Unconjugated | |
| LMNA | |
| 70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16905, 4000, 60374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction