missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A + C R453W Antibody (12A-2F5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-77401F
This item is not returnable.
View return policy
Description
Lamin A + C R453W Monoclonal specifically detects Lamin A + C R453W in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Lamin A + C R453W | |
| Monoclonal | |
| FITC | |
| NP_733821.1 | |
| LMNA | |
| Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [Accession#: NP_733821.1] | |
| 0.1 mL | |
| Cancer, Chromatin Modifiers, Chromatin Research, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers, Signal Transduction | |
| 4000 | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 12A-2F5 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Disease-associated point-mutant R453W of Lamin A + C | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction