missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lamin A/C (R482W) Monoclonal Antibody (5H8-B4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518063
This item is not returnable.
View return policy
Description
This antibody recognizes the FPLD-associated lamin A/C mutant strain R482W.
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).
Specifications
| Lamin A/C (R482W) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P02545 | |
| LMNA | |
| Lamin A Ig-fold; human (Immunogen sequence: 430-545 (6xHis-tagged fusion protein)). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 5H8-B4 | |
| Unconjugated | |
| LMNA | |
| 70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A/C; lamin A/C-like 1; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; Prelamin-A/C; PRO1; Renal carcinoma antigen NY-REN-32 | |
| Mouse | |
| Protein G | |
| RUO | |
| 4000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction