missing translation for 'onlineSavingsMsg'
Learn More

Lamin A + C Antibody (131C3), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18426047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426047 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426047 Supplier Novus Biologicals Supplier No. NBP197687MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lamin A + C Monoclonal antibody specifically detects Lamin A + C in Human, Mouse, Rat, Bovine, Canine, Hamster, Sheep samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin A + C
Applications Flow Cytometry
Classification Monoclonal
Clone 131C3
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32
Host Species Mouse
Immunogen Derived by fusion of P3/X63.Ag8.653 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with purified Rat liver lamins.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Protein Phosphatase, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 4000
Target Species Human, Mouse, Rat, Bovine, Canine, Hamster, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.