Learn More
LAMC3-Specific Mouse anti-Human, Unconjugated, Clone: 4B8C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67261-1-IG-150UL
Description
The LAMC3-Specific antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LAMC3-Specific. This antibody recognizes human, mouse antigen. The LAMC3-Specific antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| LAMC3-Specific | |
| Monoclonal | |
| 1500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, ELISA | |
| 4B8C5 | |
| Unconjugated | |
| 150 μL | |
| 10319 | |
| IgG2a |