missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda5/IGLL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Lambda5/IGLL1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235523
|
Novus Biologicals
NBP2-56135 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671688
|
Novus Biologicals
NBP2-56135-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lambda5/IGLL1 Polyclonal specifically detects Lambda5/IGLL1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Lambda5/IGLL1 | |
| Polyclonal | |
| Rabbit | |
| Immunology | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3543 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GFQSKHNSVTHVFGSGTQLTVLSQPK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 14.1, AGM2, CD179 antigen-like family member B, CD179b, CD179b antigen, Ig lambda-5, IGL1, IGL5CD179B, IGLJ14.1, IGLL, IGO, IGVPB, immunoglobulin lambda-like polypeptide 1, Immunoglobulin omega polypeptide, immunoglobulin omega polypeptide chain, immunoglobulin-related 14.1 protein, Immunoglobulin-related protein 14.1, lambda5, Pre-B lymphocyte-specific protein-2, VPREB2 | |
| IGLL1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title