missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Lambda Tint Plate
Click to view available options
For Use With (Equipment):
Motic TEC POL Microscopes
Unit Size:
Each
Description
Retards light by one full wave. For use with the following Motic™ Microscopes:
- Panthera TEC POL Binocular
- Panthera TEC POL Trinocular
- Panthera TEC POL Epi Binocular
- Panthera TEC POL Epi Trinocular
- Panthera TEC POL Digital
- Panthera TEC POL Epi Digital
- Panthera TEC POL (i)
Specifications
Specifications
| Type | Waveplate |
| Color | Black |
| For Use With (Application) | Retarding Microscope Light |
| For Use With (Equipment) | Motic TEC POL Microscopes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction