Learn More
Motic™ Lambda Plate
Click to view available options
For Use With (Equipment):
Polarizing microscope
Unit Size:
Each
Description
- Full wave plate.
- First-order red.
- 535nm. Rotatable.
Specifications
Specifications
| Type | Lambda Plate |
| For Use With (Equipment) | Polarizing microscope |