missing translation for 'onlineSavingsMsg'
Learn More

Lambda Light Chain Rat anti-Mouse, Clone: JC5-1, Novus Biologicals™

Product Code. 18395062
Change view
Click to view available options
Quantity:
1000 μg
250 μg
Unit Size:
1000µg
250µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395062 1000 μg 1000µg
18361326 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18395062 Supplier Bio-Techne Supplier No. NBP3183041000UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda Light Chain
Applications ELISA
Classification Monoclonal
Clone JC5-1
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias IGLC, immunoglobulin lambda constant group
Host Species Rat
Immunogen Mouse lambda light chain
Purification Method Protein G purified
Quantity 1000 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3537
Target Species Mouse
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.