Learn More
Lambda Light Chain Antibody (N10/2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-54421C
Product Code. 18480745
  • £444.00 / 100µL
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lambda Light Chain
Monoclonal
DyLight 650
IGLC, immunoglobulin lambda constant group
Purified IgG from human serum
100 μg
Adaptive Immunity, Immunology
3537
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
N10/2
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified