missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (LLC/1738), Alexa Fluor™ 700, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Lambda Light Chain |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | LLC/1738 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | IGLC, immunoglobulin lambda constant group |
| Host Species | Mouse |
| Immunogen | Purified human Lambda Light Chain |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?