missing translation for 'onlineSavingsMsg'
Learn More

Lambda Light Chain Antibody (LAM03) - Azide and BSA Free, Novus Biologicals™

Product Code. 18402772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402772 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402772 Supplier Novus Biologicals Supplier No. NBP2346640.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda Light Chain
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone LAM03
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 to 5 μg/mL for coating, Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 to 1 μg/mL, CyTOF-ready
Formulation 10 mM PBS
Gene Alias IGLC, immunoglobulin lambda constant group
Host Species Mouse
Immunogen Purified human lambda light chain
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3537
Target Species Human
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.