missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMA4 (Isoform 3) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10465-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody was generated against the full length (1-120aa) of isoform 3.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kDa) and three short arms (200 kDa). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.Specifications
| LAMA4 (Isoform 3) | |
| Polyclonal | |
| Unconjugated | |
| LAMA4 | |
| LAMA4 (Isoform 3), Laminin 14 subunit alpha, Laminin 8 subunit alpha, Laminin 9 subunit alpha, Laminin subunit alpha 4, laminin, alpha 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3910 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16363 | |
| LAMA4 | |
| LAMA4 (Isoform 3) Fusion Protein Ag0736 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction