missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LAIR2 Monoclonal antibody specifically detects LAIR2 in Human samples. It is validated for ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair)
Specifications
Specifications
| Antigen | LAIR2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Dilution | ELISA, ELISA Capture (Matched Antibody Pair) 50 - 200ng/well, ELISA Detection (Matched Antibody Pair) 0.1 - 0.5ug/ml |
| Gene Alias | CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634 |
| Host Species | Rabbit |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?