missing translation for 'onlineSavingsMsg'
Learn More

LAIR2 Antibody (024), Alexa Fluor™ 488 , Novus Biologicals™

Product Code. 18432708 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432708 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432708 Supplier Novus Biologicals Supplier No. NBP289685AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LAIR2 Monoclonal antibody specifically detects LAIR2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAIR2
Applications ELISA
Classification Monoclonal
Clone 024
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human LAIR2 (NP_002279.2; Met 1-Pro 152).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3904
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.