missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 (CD223) Mouse anti-Human, BV421, Clone: T47-530, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565720
This item is not returnable.
View return policy
Description
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.
Specifications
| LAG-3 (CD223) | |
| Monoclonal | |
| Brilliant Violet 421 | |
| LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
| Human LAG-3 Recombinant Protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
| IgG1 κ |
| Flow Cytometry | |
| T47-530 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction