missing translation for 'onlineSavingsMsg'
Learn More

LAG-3 Antibody (L4-PL33) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£224.00 - £499.00

Specifications

Antigen LAG-3
Clone L4-PL33
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18612664
View Documents Promotion Details
Novus Biologicals
NBP3-23442-100ug
100 μg
£499.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18621544
View Documents
Novus Biologicals
NBP3-23442-50ug
50 μg
£346.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18651334
View Documents Promotion Details
Novus Biologicals
NBP3-23442-25ug
25 ug
£224.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LAG-3 Monoclonal antibody specifically detects LAG-3 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
Specifications

Specifications

LAG-3
Monoclonal
Purified
RUO
Human, Primate
17b4, 17b4 lag3, 17b4 neutralizing, anti human lag3, Anti-LAG-3, CD223, CD223 Antigen, Human LAG3, Human LAG-3, LAG3, LAG-3, LAG-3 17b4, LAG-3 blocking, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
This LAG-3 Antibody (L4-PL33) was prepared from synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3).
Protein A or G purified
L4-PL33
Unconjugated
Mouse
Adaptive Immunity, Cancer, Immunology, Innate Immunity
PBS, 10% glycerol
3902
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.