Learn More
LAG-3 Antibody (CL14765) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26841AF700
Product Code. 30292754
  • £582.00 / 0.10mL
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAG-3
Monoclonal
Alexa Fluor 700
CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
This antibody was developed using a synthetic peptide derived from P18627, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Cancer, Immunology, Innate Immunity
3902
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry
CL14765
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified