missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 Antibody (17B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80059
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Inhibition Activity
Specifications
| LAG-3 | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 5 ug, Flow Cytometry, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Frozen 1:150, Inhibition Activity | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| Synthetic peptide corresponding to 30 aa in the N-terminus of human LAG-3. | |
| 100 μg | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
| 3902 | |
| Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Inhibition Assays | |
| 17B4 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction