missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Inhibition Activity
Specifications
Specifications
| Antigen | LAG-3 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 17B4 |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 |
| Host Species | Mouse |
| Immunogen | Synthetic peptide corresponding to 30 aa in the N-terminus of human LAG-3. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?