missing translation for 'onlineSavingsMsg'
Learn More

LAG-3 Antibody (11E3), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18421705 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421705 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421705 Supplier Novus Biologicals Supplier No. NBP197662AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LAG-3 Monoclonal antibody specifically detects LAG-3 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAG-3
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 11E3
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
Host Species Mouse
Immunogen This LAG-3 Antibody (11E3) was prepared from recombinant human LAG-3 (lymphocyte activation gene-3).
Purification Method Protein G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.