missing translation for 'onlineSavingsMsg'
Learn More

Lafutidine, MedChemExpress

Product Code. 16448368
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16448368

Brand: Medchem Express HYB0160

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Lafutidine (FRG-8813) is a histamine H2-receptor antagonist (H2RA), with proven gastric mucosal protective effects. Lafutidine can be used for the research of gastroesophageal reflux disease.

Lafutidine (FRG-8813) is a histamine H2-receptor antagonist (H2RA), with proven gastric mucosal protective effects. Lafutidine can be used for the research of gastroesophageal reflux disease.

Specifications

Chemical Name or Material Lafutidine
Physical Form Solid
CAS 118288-08-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H29N3O4S
For Use With (Application) Neuroscience-Neuromodulation
Synonym FRG-8813
Solubility Information DMSO : ≥ 50 mg/mL (115.86 mM)
SMILES O=C(NC/C=C\COC1=NC=CC(CN2CCCCC2)=C1)CS(CC3=CC=CO3)=O
Molecular Weight (g/mol) 431.55
Formula Weight 431.55
Percent Purity 98.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.