missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565781
This item is not returnable.
View return policy
Description
Immunogen sequence: KSQGEKDSSSR LATSTSNTLS ANHCNMNINS VAIPINKNEK MLRSPISPLS DASKHKYTSE DLTSSSRPNG NSLFTSASSS KKPKADSQLQ PHGGDLTKAA HNNSENIPLH KSRPQTKPWS PGSNGHRDCK RQKLVFDDMP Highest antigen sequence identity to the following orthologs - mouse 79%, rat 76%.
This gene encodes a tissue-restricted nuclear transcriptional activator that is preferentially expressed in lymphoid tissue. Isolation of this protein initially defined a highly conserved LAF4/MLLT2 gene family of nuclear transcription factors that may function in lymphoid development and oncogenesis. In some ALL patients, this gene has been found fused to the gene for MLL. Multiple alternatively spliced transcript variants that encode different proteins have been found for this gene.
Specifications
| LAF4 | |
| Polyclonal | |
| Unconjugated | |
| AFF3 | |
| 3222402O04Rik; A730046J16; AF4/FMR2 family member 3; AF4/FMR2 family, member 3; AFF3; id:ibd5053; LAF4; LAF-4; lymphoid nuclear protein 4; lymphoid nuclear protein related to AF4; MLLT2-like; MLLT2-related protein; protein LAF-4; si:dkey-19j7.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51826 | |
| AFF3 | |
| Recombinant Human LAF4. Recombinant protein control fragment (Product #RP-106460). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction