missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Laemmli SDS Sample Buffer with Pyronin Y, reducing, 4X

Product Code. 15480227
Change view
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480227 50 mL 50mL
15470227 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480227 Supplier Thermo Scientific™ Supplier No. J62115.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For protein sample preparation to be used in the Laemmli SDS-PAGE system

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Pink
Concentration 4X
Form Liquid
For Use With (Application) Laemmli SDS-PAGE
Quantity 50 mL
Product Type Sample Loading Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.