missing translation for 'onlineSavingsMsg'
Learn More

Laduviglusib, MedChemExpress

Product Code. 16456317
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456317 5 mg 5mg
16419996 50 mg 50mg
16041282 10 mg 10mg
16414696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16456317 Supplier Medchem Express Supplier No. HY10182
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Laduviglusib (CHIR-99021) is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM. Laduviglusib shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib enhances mouse and human embryonic stem cells self-renewal. Laduviglusib induces autophagy.

Laduviglusib (CHIR-99021) is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM. Laduviglusib shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib enhances mouse and human embryonic stem cells self-renewal. Laduviglusib induces autophagy.

Specifications

Chemical Name or Material Laduviglusib
CAS 252917-06-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H18Cl2N8
Quantity 5 mg
Synonym CHIR-99021 CT99021
Solubility Information DMSO : 16.67 mg/mL (35.82 mM; Need ultrasonic)
SMILES N#CC1=CC=C(N=C1)NCCNC2=NC=C(C(C3=CC=C(Cl)C=C3Cl)=N2)C4=NC=C(N4)C
Molecular Weight (g/mol) 465.34
Formula Weight 465.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.