Learn More
Toesaver™ Ladies Black Microfiber Lace Safety Shoe PU Sole
Click to view available options
Size:
2
3
4
5
6
7
8
Description
- 200 Joule toecap protection
- Heat resistant to 200°C
- SRA - outsole slip resistance
- EN ISO 20345-S1
Specifications
Specifications
| Color | Black |
| Material | Microfiber |
| Size | 2 |
| Sole Material | Polyurethane |