missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAD1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145485
This item is not returnable.
View return policy
Description
Anchoring filament protein which is a component of the basement membrane zone.
Specifications
| LAD1 | |
| Polyclonal | |
| Unconjugated | |
| LAD1 | |
| LAD; Lad1; Lad-1; LADA; ladinin; ladinin 1; ladinin-1; linear IgA bullous dermatosis antigen; Linear IgA disease antigen; Linear IgA disease antigen homolog | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 16763, 313325 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P57016 | |
| LAD1 | |
| Recombinant Rat LAD1 protein, Met1-Pro140 (Accession #G3V779). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction