missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lactulose, 99%

Product Code. 15462979
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
This item is not returnable. View return policy

Product Code. 15462979

Brand: Thermo Scientific Chemicals J60160.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A synthetic sugar | CAS: 4618-18-2 | C12H22O11 | 342.297 g/mol

It is not absorbed in the gut and is frequently used in the treatment of constipation and hepatic encephalopathy. actulose is also a food ingredient, better known as galactofructose, with sweet taste and offering beneficial health benefits on digestive health. Also used as a test of small intestine bacterial overgrowth (SIBO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (76.4%) at 30°C, water (81%) at 60°C, and water (>86%) at 90°C and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4618-18-2
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00151469
InChI Key JCQLYHFGKNRPGE-DJYZFUSFSA-N
Synonym lactulose
PubChem CID 131839611
IUPAC Name (2S,3S,4S,5R,6R)-2-[(2R,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C2O)(CO)O)CO)O)O)O)O

Specifications

Melting Point 169°C
Color White
Density 1.32
Quantity 50 g
Beilstein 93773
Merck Index 14,5346
Solubility Information Soluble in water (76.4%) at 30°C,water (81%) at 60°C,and water (>86%) at 90°C and methanol.
Formula Weight 342.3
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Lactulose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.