missing translation for 'onlineSavingsMsg'
Learn More

Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™

Product Code. 10315810 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10315810

Brand: Fisher Chemical L/0250/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lactose Monohydrate, Certified AR for Analysis, Fine Powder

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
MDL Number 71587
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686

Specifications

Melting Point 219°C
Color White
Packaging Plastic powder jar
Quantity 500 g
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Molecular Weight (g/mol) 360.312
Formula Weight 360.3g/mol
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.