missing translation for 'onlineSavingsMsg'
Learn More

Lactose, MedChemExpress

Product Code. 16402379
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402379 Supplier Medchem Express Supplier No. HYB2123
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lactose, a major sugar in the milk of most species, could regulate human’s intestinal microflora.

Lactose, a major sugar in the milk of most species, could regulate human’s intestinal microflora.

Specifications

Chemical Name or Material Lactose
CAS 63-42-3
Color White
Purity Grade Notes Research
Molecular Formula C12H22O11
Quantity 1 g
Solubility Information H2O : 100 mg/mL (292.14 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (146.07 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](O)[C@H](O)[C@@H](O)[C@H](O[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)CO)O1
Molecular Weight (g/mol) 342.3
Formula Weight 342.3
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.