missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Lactoperoxidase (Bovine)

Product Code. 16151750
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16151750

Brand: Abnova™ P5202.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 77.5kDa
Name Lactoperoxidase (Bovine)
Preparation Method Native protein purified from Bovine Milk
Quantity 10 mg
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name LACTOPEROXIDASE
Assays Lactoperoxidase Assay
Biological Activity ≥35 units per mg dry weight. One Unit reduces one micromole of hydrogen peroxide per minute at 25°C, pH 6.0.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Milk
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.