missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoperoxidase Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-61572FR
This item is not returnable.
View return policy
Description
Lactoperoxidase Polyclonal antibody specifically detects Lactoperoxidase in Human, Bovine, Primate samples. It is validated for Western Blot
Specifications
| Lactoperoxidase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Bovine, Primate | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
| Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot# P22079] | |
| 0.1 mL | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 4025 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction