missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoperoxidase Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-61574AF532
This item is not returnable.
View return policy
Description
Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Mouse, Bovine, Goat, Sheep samples. It is validated for Western Blot.
Specifications
| Lactoperoxidase | |
| Polyclonal | |
| Western Blot | |
| EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 4025.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LPO | |
| Synthetic peptide made to a C-terminal portion of the human Lactoperoxidase protein (within residues 650-712). [Swiss-ProtNo. P22079] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Bovine, Goat, Sheep | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction