missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (SR2256), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £351.00
Specifications
| Antigen | Lactoferrin |
|---|---|
| Clone | SR2256 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630194
|
Novus Biologicals
NBP3-22072-100ul |
100 μg |
£351.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606824
|
Novus Biologicals
NBP3-22072-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| Lactoferrin | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| A synthesized peptide derived from human Lactoferrin (Uniprot #: P02788) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2256 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4057 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title