missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (LTF/4082) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14113JF635
This item is not returnable.
View return policy
Description
Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| Lactoferrin | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| Recombinant fragment (around aa614-645) of human Lactoferrin protein (exact sequence is proprietary) (Uniprot: IDP02788) | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules | |
| 4057 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| LTF/4082 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction