missing translation for 'onlineSavingsMsg'
Learn More

Lactoferrin Antibody (002), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18362711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362711 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362711 Supplier Novus Biologicals Supplier No. NBP290704AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lactoferrin Monoclonal specifically detects Lactoferrin in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactoferrin
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Lactoferrin (AAH06904.1; Met1-Gln707).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules
Primary or Secondary Primary
Gene ID (Entrez) 4057
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.