missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferricin Antibody (5F12.1.2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11195
This item is not returnable.
View return policy
Description
Lactoferricin Monoclonal specifically detects Lactoferricin in Bovine samples. It is validated for Western Blot, Immunoassay.
Specifications
| Lactoferricin | |
| Monoclonal | |
| 0.1 mg/ml | |
| Western Blot, Immunoassay | |
| Bovine Lactoferricin, Lactoferricin B, LF, LF-B, Lfcin, Lfcin B | |
| Monoclonal antibody clone 5F12.1.2, anti bovine Lactoferricin B is highly specific for bovine Lactoferricin B. | |
| 50 μg | |
| Primary | |
| Bovine | |
| Purified |
| Western Blot, Immunoassay | |
| 5F12.1.2 | |
| Unconjugated | |
| 0.2 um filtered solution in PBS, 0.1% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4057 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction