missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactobionic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Lactobionic acid |
| CAS | 96-82-2 |
| Molecular Formula | C12 H22 O12 |
| Quantity | 10 g |
| Synonym | D-Gluconic acid, 4-O-β-D-galactopyranosyl-, Lactobionic acid (7CI,8CI), Lactonic acid (6CI), 4-O-β-D-Galactopyranosyl-D-gluconic acid, 4-(β-D-Galactosido)-D-gluconic acid, 4-O-β-Galactopyranosyl-D-gluconic acid, D-Lactobionic acid, Galactosylgluconic acid |
| InChI Formula | InChI=1S/C12H22O12/c13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12/h3-10,12-20H,1-2H2,(H,21,22)/t3-,4-,5+,6+,7-,8-,9-,10-,12+/m1/s1 |
| SMILES | OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(=O)O |
| IUPAC Name | (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanoic acid |
| Molecular Weight (g/mol) | 358.3 |
| Formula Weight | 358.1111 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction