missing translation for 'onlineSavingsMsg'
Learn More

Lactobionic acid, 97%

Product Code. 15457699
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
This item is not returnable. View return policy

Product Code. 15457699

Brand: Thermo Scientific Alfa Aesar J66368.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-82-2 | C12H22O12 | 358.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-82-2
Molecular Formula C12H22O12
Molecular Weight (g/mol) 358.296
MDL Number MFCD00078147
InChI Key JYTUSYBCFIZPBE-AMTLMPIISA-N
Synonym lactobionic acid, 4-o-beta-d-galactopyranosyl-d-gluconic acid, unii-65r938s4dv, 4-beta-d-galactosido-d-gluconic acid, dsstox_cid_28787, dsstox_rid_83056, dsstox_gsid_48861, lactobionicacid, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-hexanoic acid, prestwick_560
PubChem CID 7314
ChEBI CHEBI:55481
IUPAC Name (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanoic acid
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(C(=O)O)O)O)O)O)O)O

Specifications

Melting Point 113°C to 118°C
Color White
Odor Odorless
Quantity 250 g
Merck Index 14,5342
Solubility Information Soluble in water
Optical Rotation +25° (c=10 in Water)
Formula Weight 358.3
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lactobionic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.