missing translation for 'onlineSavingsMsg'
Learn More

Lactitol Monohydrate, TRC

Product Code. 30483267
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity unitSize
30483267 5 mg 5mg
1 options
This item is not returnable. View return policy

Product Code. 30483267

Brand: Toronto Research Chemicals TRCL0691255MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Lactitol monohydrate
CAS 81025-04-9
Molecular Formula C12 H24 O11 . H2 O
Quantity 5 mg
Synonym Lactitol monohydrate, 4-O-(β-d-Galactopyranosyl)-d-glucitol monohydrate
InChI Formula InChI=1S/C12H24O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h4-21H,1-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1
SMILES O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)CO
IUPAC Name (2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate
Molecular Weight (g/mol) 362.33
Formula Weight 362.1424
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.