missing translation for 'onlineSavingsMsg'
Learn More

Lactitol monohydrate, MedChemExpress

Product Code. 16488746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
16488746 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 16488746

Brand: Medchem Express HYB1389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lactitol monohydrate is a disaccharide analogue of lactulose.

Lactitol monohydrate is a disaccharide analogue of lactulose.

Specifications

Chemical Name or Material Lactitol monohydrate
CAS 81025-04-9
Color White
Purity Grade Notes Research
Molecular Formula C12H26O12
Quantity 100 mg
Synonym D-Lactitol monohydrate
Solubility Information H2O : 120 mg/mL (331.19 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (275.99 mM; Need ultrasonic)
SMILES OC[C@@H]([C@H]([C@@H]([C@@H](CO)O)O[C@H]1[C@@H]([C@H]([C@H]([C@@H](CO)O1)O)O)O)O)O.[H]O[H]
Molecular Weight (g/mol) 362.33
Formula Weight 362.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.