missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, lithium salt, 99%, extra pure

Product Code. 10390674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10390674 500 g Glass bottle 500g
10485651 5 g Glass bottle 5g
10301401 25 g Glass bottle 25g
10219433 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10390674 Supplier Thermo Scientific Supplier No. 413335000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 867-55-0 | C3H5LiO3 | 96.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-55-0
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065399
InChI Key GKQWYZBANWAFMQ-UHFFFAOYSA-M
Synonym lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate
PubChem CID 23673452
IUPAC Name lithium;2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Absorbance 10% aq. soln., 10% aq. soln. at 0nm, 0.07 max. at 260nm
Color White
pH 6 to 9 (1% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula CH3CH(OH)CO2Li
Quantity 500 g
Beilstein 03, IV, 633
Formula Weight 96.01
Percent Purity ≥98.5%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Lactic acid, lithium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.