missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, Extra Pure, SLR, Fisher Chemical™
£74.95 - £327.00
Chemical Identifiers
CAS | 79-33-4 |
---|---|
Molecular Formula | C3H6O3 |
Molecular Weight (g/mol) | 90.08 |
MDL Number | MFCD00004520 |
InChI Key | JVTAAEKCZFNVCJ-UHFFFAOYNA-N |
Synonym | lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum |
PubChem CID | 612 |
ChEBI | CHEBI:78320 |
IUPAC Name | 2-hydroxypropanoic acid |
SMILES | CC(O)C(O)=O |
Description
Chemical Identifiers
79-33-4 | |
90.08 | |
JVTAAEKCZFNVCJ-UHFFFAOYNA-N | |
612 | |
2-hydroxypropanoic acid |
C3H6O3 | |
MFCD00004520 | |
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum | |
CHEBI:78320 | |
CC(O)C(O)=O |
Specifications
1,2 | |
125°C | |
C3H6O3 | |
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum | |
CC(O)C(O)=O | |
90.08 | |
CHEBI:78320 | |
HDPE plastic bottle |
79-33-4 | |
Yellow | |
MFCD00004520 | |
JVTAAEKCZFNVCJ-UHFFFAOYNA-N | |
2-hydroxypropanoic acid | |
612 | |
90.08g/mol | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- L(+)-Lactic acid, 90% solution in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.