missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, ACS, 85.0-90.0% aq. soln.

Product Code. 11347878
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347878 2 kg 2kg
11327878 100 g 100g
11337878 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347878 Supplier Thermo Scientific Alfa Aesar Supplier No. 036415.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol

It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Melting Point 18°C
Boiling Point 122°C (15mmHg)
Color Colorless to Yellow
Assay Percent Range 85.0 to 90.0%
Beilstein 1209341
Merck Index 14,5336
Solubility Information Soluble in water,ethanol,furfurol,alcohol and water. Partially soluble in diethyl ether. Practically insoluble in chloroform,petroleum ether,carbon disulfide.
Concentration 85.0 to 90.0% aq. soln.
Formula Weight 90.08
Grade ACS
Density 1.209 g/mL
Quantity 2 kg
Flash Point >110°C (230°F)
Physical Form Liquid
Refractive Index 1.427
Chemical Name or Material Lactic acid, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.