missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Lactic Acid 10%

Product Code. 10076732
Click to view available options
Quantity:
10 x 1 mL
10 x 5 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10076732

Brand: Thermo Scientific™ SR0021H

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A solution for addition to culture media, primarily to lower the pH

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Country of Origin UK
Format Box
Media Type Additive
Product Type Media Supplement
For Use With (Application) Lowers pH of Culture Media
Packaging Type 10 x 1 mL
Quantity 10 x 1 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • LACTIC ACID 10%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Lactic Acid 90%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.