missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | LACTB |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384773
|
Bio-Techne
NBP3-16652-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18365656
|
Novus Biologicals
NBP3-16652-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LACTB Polyclonal antibody specifically detects LACTB in Human, Mouse samples. It is validated for Western BlotSpecifications
| LACTB | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| AmpC, EC 3.4, FLJ14902, G24, lactamase, beta, mitochondrial 39S ribosomal protein L56, mitochondrial ribosomal protein L56, MRPL56, serine beta lactamase-like protein LACTB, serine beta-lactamase-like protein LACTB, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-290 of human LACTB (NP_116246.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| PBS, 50% glycerol, pH7.3 | |
| 114294 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title