missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-53421SS
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase B Polyclonal specifically detects Lactate Dehydrogenase B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| Lactate Dehydrogenase B | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LDHB | |
| Partial recombinant human Lactate Dehydrogenase B protein (amino acids 1-93) [P07195] | |
| 0.025 mg | |
| HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway | |
| 3945 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1 ug/ml, Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:400, Knockout Validated | |
| EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction